Learn More
ABclonal Technology Entrans 2X qPCR Probe Master Mix with UDG
Supplier: ABclonal Technology RK21222250RXN
Entrans 2X qPCR Probe Master Mix with UDG is aready-to-use reagent for probe-based qPCRreactions, containing all components exceptprimers, probes and templates. This master mixincludes a chemically modified version of Taqpolymerase with Hot-start capability to inhibit nonspecific amplification, while allowing high efficiency,sensitivity and reproducibility in qPCR amplification.At the same time, and has joined the UDG antipollution system. The optimization of the Buffersystem allows the product to perform multiplefluorescence quantitative experiments, andIt is suitable for multiple species and provides apowerful tool for multi-disciplinary experimentalneeds.