Learn More
ABclonal Technology MethyLight 2X qPCR Probe Master Mix
SDP
Supplier:  ABclonal Technology RK212405ML
Encompass_Preferred
MethyLight 2X qPCR Probe Master Mix is a probe-based reagent for real time PCR reaction. It is a powerful tool for MSP analysis of methylated DNA and multi-target detection in epigenetic research.It is an optimized premix containing DNA polymerase, dNTPs, MgCl2, KCI and other stabilizers. The optimized master mixes ensure that the PCR products in a reaction with bisulfite converted DNA as starting material are amplified with high efficiency and sensitivity. This product is a 2× concentration master mix, which contains all the components required for MethyLight qPCR reactions except primers, probes and templates.
Catalog No. 50-001-38554