Learn More
ABclonal Technology Nt.BspQI
SDP
Catalog No. 5000137778
Encompass_Preferred
5…GCTCTTCN...3 3…CGAGAAGN...5
Catalog No. 50-001-37778 Supplier ABclonal Technology Supplier No. RK213005000U
5…GCTCTTCN...3 3…CGAGAAGN...5