Learn More
ABclonal Technology RNase Inhibitor, Mammalian
SDP
Catalog No. 5000135153
Encompass_Preferred
RNase Inhibitor, Mammalian specifically inhibits RNases A, B and C. It inhibits RNases by binding noncovalently in a 1:1 ratio with high affinity. It is not effective against RNase 1, RNase T1, S1 Nuclease, RNase H or RNase from Aspergillus. In addition, no inhibition of polymerase activity is observed when RNase Inhibitor is used with Taq DNA Polymerase, AMV or M-MuLV Reverse Transcriptases, or Phage RNA Polymerases (SP6, T7, or T3). The 50 kDa protein inhibits RNases by binding noncovalently in a 1:1 ratio with an association constant greater than 1014.
Catalog No. 50-001-35153 Supplier ABclonal Technology Supplier No. RK2140110000U
RNase Inhibitor, Mammalian specifically inhibits RNases A, B and C. It inhibits RNases by binding noncovalently in a 1:1 ratio with high affinity. It is not effective against RNase 1, RNase T1, S1 Nuclease, RNase H or RNase from Aspergillus. In addition, no inhibition of polymerase activity is observed when RNase Inhibitor is used with Taq DNA Polymerase, AMV or M-MuLV Reverse Transcriptases, or Phage RNA Polymerases (SP6, T7, or T3). The 50 kDa protein inhibits RNases by binding noncovalently in a 1:1 ratio with an association constant greater than 1014.