Learn More
ABclonal Technology High Salt-T4 DNA Ligase
SDP
Supplier:  ABclonal Technology RK21506100000U
Encompass_Preferred
High Salt-T4 DNA Ligase is a salt-tolerant mutant of T4 DNA Ligase that catalyzes the formation of a phosphodiester bond between juxtaposed 5? phosphate and 3? hydroxyl termini in duplex DNA or RNA. It is designed to function at higher salt concentrations than wild type T4 DNA Ligase. The catalytic reaction requires ATP as a cofactor. High Salt-T4 DNA Ligase will join blunt end and cohesive end termini at salt concentrations as high as 300 mM without any loss in activity, and can also repair single-stranded nicks in duplex DNA, RNA or DNA/RNA hybrids. At the same time, T4 DNA ligase closes the gaps in these DNA substrates.
Catalog No. 50-001-38049