Learn More
ABclonal Technology RK21623-5uleach Unique Dual I
SDP
Catalog No. 5000135048
Encompass_Preferred
ABclonal s Unique Dual Index for Illumina products contain universal truncated adapters and Unique Dual Index (UDI) primers for sample preparation for Next Generation Sequencing on Illumina sequencing The MidiSet (CAT NO RK21623) provides 24 unique primer pairs (UDI Primers 1-24) All sets include 15 M truncated adapters along with the listed primers The sample index length is 8 nucleotides
Catalog No. 50-001-35048 Supplier ABclonal Technology Supplier No. RK216235ULEACH
ABclonal s Unique Dual Index for Illumina products contain universal truncated adapters and Unique Dual Index (UDI) primers for sample preparation for Next Generation Sequencing on Illumina sequencing The MidiSet (CAT NO RK21623) provides 24 unique primer pairs (UDI Primers 1-24) All sets include 15 M truncated adapters along with the listed primers The sample index length is 8 nucleotides