Learn More
ABclonal Technology RK21626-5uleach Unique Dual I
SDP
Catalog No. 5000135198
Encompass_Preferred
ABclonal s Unique Dual Index for Illumina products contain universal truncated adapters and Unique Dual Index (UDI) primers for sample preparation for Next Generation Sequencing on Illumina sequencing Set C (CAT NO RK21626) provides 48 unique primer pairs (UDI Primers 97-144) All sets include 15 M truncated adapters along with the listed primers The sample index length is 8 nucleotides
Catalog No. 50-001-35198 Supplier ABclonal Technology Supplier No. RK216265ULEACH
ABclonal s Unique Dual Index for Illumina products contain universal truncated adapters and Unique Dual Index (UDI) primers for sample preparation for Next Generation Sequencing on Illumina sequencing Set C (CAT NO RK21626) provides 48 unique primer pairs (UDI Primers 97-144) All sets include 15 M truncated adapters along with the listed primers The sample index length is 8 nucleotides