Learn More
ABclonal Technology RK21687-5uleach DNA Adapter U
SDP
Catalog No. 5000138653
Encompass_Preferred
The Truncated DNA Adapter (UDI) Kit for MGI kits contain Unique Dual Index adapters designed for the MGI sequencing platform Using these products to construct libraries can result in libraries with dual unique indexes The truncated adapters offer higher ligation efficiency fewer adapter dimers and higher library yield The adapters in these kits are truncated and cannot be used for PCR-free library construction It is necessary to use MGI UDI Primers for at least 2-3 cycles to obtain a structurally complete library
Catalog No. 50-001-38653 Supplier ABclonal Technology Supplier No. RK216875ULEACH
The Truncated DNA Adapter (UDI) Kit for MGI kits contain Unique Dual Index adapters designed for the MGI sequencing platform Using these products to construct libraries can result in libraries with dual unique indexes The truncated adapters offer higher ligation efficiency fewer adapter dimers and higher library yield The adapters in these kits are truncated and cannot be used for PCR-free library construction It is necessary to use MGI UDI Primers for at least 2-3 cycles to obtain a structurally complete library