Learn More
ABclonal Technology RK21706-25uleach SetA UDI 1-
SDP
Catalog No. 5000138046
Encompass_Preferred
The Unique Dual Index Adapter Plate for Illumina contains a total of 384 UDI adapters UDI adapters have indexes on both ends of P5 and P7 and the indices on P5 and P7 are uniquely paired The UDI Adapter is available in 96-well plates of 25 L per well for automated workstations This product is suitable for simultaneous sequencing of a large number of samples and the UDI Adapter concentration is 15 M This kit does not contain amplification primers common to the Illumina platform which need to bepurchased separately or matched by yourself
Catalog No. 50-001-38046 Supplier ABclonal Technology Supplier No. RK2170625ULEACH
The Unique Dual Index Adapter Plate for Illumina contains a total of 384 UDI adapters UDI adapters have indexes on both ends of P5 and P7 and the indices on P5 and P7 are uniquely paired The UDI Adapter is available in 96-well plates of 25 L per well for automated workstations This product is suitable for simultaneous sequencing of a large number of samples and the UDI Adapter concentration is 15 M This kit does not contain amplification primers common to the Illumina platform which need to bepurchased separately or matched by yourself