Learn More
ABclonal Technology RK21708-25uleach SetC UDI 19
SDP
Catalog No. 5000138558
Encompass_Preferred
Unique Dual Index Adapter Plate for Illumina Set A/B/C/D contains a total of 384 UDI Adapters The UDI Adapters have indexes located at both the P5 and P7 ends with the indexes on the P5 and P7 ends being unique The concentration of the UDI Adapters is 15 M aliquoted into a 96-well plate with 25 L per well suitable for automated workstations This product is designed for the simultaneous sequencing of a large number of samples Universal amplification primers are not included in these kits they can typically be found in ABclonals library preparation kits
Catalog No. 50-001-38558 Supplier ABclonal Technology Supplier No. RK2170825ULEACH
Unique Dual Index Adapter Plate for Illumina Set A/B/C/D contains a total of 384 UDI Adapters The UDI Adapters have indexes located at both the P5 and P7 ends with the indexes on the P5 and P7 ends being unique The concentration of the UDI Adapters is 15 M aliquoted into a 96-well plate with 25 L per well suitable for automated workstations This product is designed for the simultaneous sequencing of a large number of samples Universal amplification primers are not included in these kits they can typically be found in ABclonals library preparation kits