Learn More
ABclonal Technology RK21724-25uleach Set D UDI Pr
SDP
Catalog No. 5000138556
Encompass_Preferred
ABclonal Truncated DNA Adapter (UDI) Plate for MGI contains universal truncated adapters and Unique Dual Index (UDI) primers for sample preparation for Next Generation Sequencing on MGI platform The Set A provides 96 unique primer pairs (MGI UDI Primers 1-96) the Set B provides 96 unique primer pairs (MGI UDI Primers 97-192) Set C provides 96 unique primer pairs (MGI UDI Primers 193-288) Set B provides 96 unique primer pairs (MGI UDI Primers 289-384) The kit can not be used to perform PCR-Free DNA lib preparation and 2-3 cycles of amplification are necessary The kit does not contain MGI Truncated Adapter A7(ABclonal CAT NO RK21720)
Catalog No. 50-001-38556 Supplier ABclonal Technology Supplier No. RK2172425ULEACH
ABclonal Truncated DNA Adapter (UDI) Plate for MGI contains universal truncated adapters and Unique Dual Index (UDI) primers for sample preparation for Next Generation Sequencing on MGI platform The Set A provides 96 unique primer pairs (MGI UDI Primers 1-96) the Set B provides 96 unique primer pairs (MGI UDI Primers 97-192) Set C provides 96 unique primer pairs (MGI UDI Primers 193-288) Set B provides 96 unique primer pairs (MGI UDI Primers 289-384) The kit can not be used to perform PCR-Free DNA lib preparation and 2-3 cycles of amplification are necessary The kit does not contain MGI Truncated Adapter A7(ABclonal CAT NO RK21720)