Learn More
ABclonal Technology AFTSpin Multifunction DNA Purification Kit
SDP
Catalog No. 5000138043
Encompass_Preferred
The AFTSpin Multifunction DNA Purification Kit enables rapid gel recovery and PCR product purification using silica-column technology. DNA binds to the membrane in high-salt conditions, while impurities are removed via wash steps. A unique gel lysis/binding solution (yellow for easy pH monitoring) combines gel digestion and DNA binding, streamlining agarose recovery, PCR cleanup, and enzyme digestion purification. Delivers high-purity DNA with improved efficiency in a single kit.
Catalog No. 50-001-38043 Supplier ABclonal Technology Supplier No. RK3010050RXN
The AFTSpin Multifunction DNA Purification Kit enables rapid gel recovery and PCR product purification using silica-column technology. DNA binds to the membrane in high-salt conditions, while impurities are removed via wash steps. A unique gel lysis/binding solution (yellow for easy pH monitoring) combines gel digestion and DNA binding, streamlining agarose recovery, PCR cleanup, and enzyme digestion purification. Delivers high-purity DNA with improved efficiency in a single kit.