Learn More
ABclonal Technology AFTSpin EndoFree Plasmid Mini Kit
SDP
Catalog No. 5000135196
Encompass_Preferred
This kit uses a modified SDS-alkaline lysis method to lyse cells. Crude extract is selectively bound to the spin column by a unique endotoxin scavenger and centrifuged to remove endotoxin. The silicone matrix membrane in the spin column selectively binds plasmid DNA at high salt and low pH conditions, allowing for impurities and other bacterial components to be removed by Buffer PR and Buffer WB. The purified plasmid DNA is eluted from the silicone matrix membrane using low-salt and high-pH Buffer EB.
Catalog No. 50-001-35196 Supplier ABclonal Technology Supplier No. RK3010250RXN
This kit uses a modified SDS-alkaline lysis method to lyse cells. Crude extract is selectively bound to the spin column by a unique endotoxin scavenger and centrifuged to remove endotoxin. The silicone matrix membrane in the spin column selectively binds plasmid DNA at high salt and low pH conditions, allowing for impurities and other bacterial components to be removed by Buffer PR and Buffer WB. The purified plasmid DNA is eluted from the silicone matrix membrane using low-salt and high-pH Buffer EB.