Learn More
ABclonal Technology AFTSpin EndoFree Plasmid Midi Kit
SDP
Catalog No. 5000138656
Encompass_Preferred
This kit uses a modified SDS-alkaline lysis method to lyse cells. The special Buffer P4 and the Filtration Columns 1 can effectively remove impurities such as endotoxin, protein and other impurities.The kit is suitable for extracting plasmid DNA from 5-15 mL bacterial cultures, and the whole experimental procedure of plasmid DNA extraction could be finished within 1 h. Plasmid DNA prepared is suitable for a variety of downstream applications including restriction enzyme digestion, PCR, sequencing, ligation, transformation and cell transfection.
Catalog No. 50-001-38656 Supplier ABclonal Technology Supplier No. RK3010550RXN
This kit uses a modified SDS-alkaline lysis method to lyse cells. The special Buffer P4 and the Filtration Columns 1 can effectively remove impurities such as endotoxin, protein and other impurities.The kit is suitable for extracting plasmid DNA from 5-15 mL bacterial cultures, and the whole experimental procedure of plasmid DNA extraction could be finished within 1 h. Plasmid DNA prepared is suitable for a variety of downstream applications including restriction enzyme digestion, PCR, sequencing, ligation, transformation and cell transfection.