Learn More
ABclonal Technology AFTSpin Universal Plant Fast DNA Extraction Kit
SDP
Catalog No. 5000135314
Encompass_Preferred
This kit is suitable for rapid and simple extraction of genomic DNA from various fresh plant tissues or cultured plant cells. The extracted genomic DNA fragments are large with high purity and stability and can be directly used for PCR, digestion, hybridization and others. The extraction process does not require phenol/chloroform extraction.
Catalog No. 50-001-35314 Supplier ABclonal Technology Supplier No. RK3011150RXN
This kit is suitable for rapid and simple extraction of genomic DNA from various fresh plant tissues or cultured plant cells. The extracted genomic DNA fragments are large with high purity and stability and can be directly used for PCR, digestion, hybridization and others. The extraction process does not require phenol/chloroform extraction.