Learn More
ABclonal Technology AFTSpin Tissue/Cell Fast RNA Extraction Kit for Animal
SDP
Catalog No. 5000136634
Encompass_Preferred
This kit does not rely on toxic reagents such as phenol and chloroform, and can be used for rapid RNA extraction from common animal tissue and cell samples. The excellent silica gel column purification technology can effectively remove residual gDNA and quickly extract animal total RNA. Extracted RNA can be directly used in reverse transcription PCR, qPCR and RNA library construction experiments.
Catalog No. 50-001-36634 Supplier ABclonal Technology Supplier No. RK3012050RXN
This kit does not rely on toxic reagents such as phenol and chloroform, and can be used for rapid RNA extraction from common animal tissue and cell samples. The excellent silica gel column purification technology can effectively remove residual gDNA and quickly extract animal total RNA. Extracted RNA can be directly used in reverse transcription PCR, qPCR and RNA library construction experiments.