Learn More
ABclonal Technology AFTSpin Complex Plant Fast RNA Extraction Kit
SDP
Catalog No. 5000138654
Encompass_Preferred
This kit is used for RNA extraction from complex plants (polysaccharide&polyphenol-rich plants). The kit does not rely on toxic reagents such as phenol and chloroform, and can remove residual gDNA by silica gel column purification technology and quickly extract total plant RNA. Extracted RNA can be directly used in RT-PCR, qPCR and RNA library construction experiments.
Catalog No. 50-001-38654 Supplier ABclonal Technology Supplier No. RK3012250RXN
This kit is used for RNA extraction from complex plants (polysaccharide&polyphenol-rich plants). The kit does not rely on toxic reagents such as phenol and chloroform, and can remove residual gDNA by silica gel column purification technology and quickly extract total plant RNA. Extracted RNA can be directly used in RT-PCR, qPCR and RNA library construction experiments.