Learn More
ABclonal Technology AFTSpin Tissue/Cell Fast miRNA Extraction Kit
SDP
Catalog No. 5000135313
Encompass_Preferred
This kit is suitable for the rapid extraction of microRNA and other small RNAs from various cells and tissues. It uses a unique lysis solution to rapidly lyse cells and inactivate cellular RNases. Organic extraction is employed to remove proteins and DNA. The special silicon - based matrix membrane in the spin column can adsorb total RNA, microRNA, and other various small RNAs. Subsequently, through a series of rapid washing-centrifugation steps, cell metabolites, proteins, and other impurities are further removed. Finally, a low - salt elution buffer is used to elute pure RNA from the silicon - based matrix membrane.
Catalog No. 50-001-35313 Supplier ABclonal Technology Supplier No. RK3012650RXN
This kit is suitable for the rapid extraction of microRNA and other small RNAs from various cells and tissues. It uses a unique lysis solution to rapidly lyse cells and inactivate cellular RNases. Organic extraction is employed to remove proteins and DNA. The special silicon - based matrix membrane in the spin column can adsorb total RNA, microRNA, and other various small RNAs. Subsequently, through a series of rapid washing-centrifugation steps, cell metabolites, proteins, and other impurities are further removed. Finally, a low - salt elution buffer is used to elute pure RNA from the silicon - based matrix membrane.