Learn More
ABclonal Technology ABScript miRNA First-Strand Synthesis Kit (by tailing A)
SDP
Catalog No. 5000137775
Encompass_Preferred
This kit is suitable for cDNA first strand synthesis using microRNA as template through the tail addition method, where the Poly (A) tail addition reaction and reverse transcription reaction at the 3 end of miRNA can be efficiently carried out simultaneously. ABScript miRNA-A Enzyme Mix contains Poly (A) Polymerase (PAP) and reverse transcriptase. PAP is mainly used to add Poly (A) tails at the 3 end of RNA molecules, and can also specifically recognize single stranded RNA, effectively avoiding RT reactions of pre-miRNA with double stranded or stem-loop structures. The modified reverse transcriptase lacks of RNase H activity and increases its affinity with RNA, resulting in a significant improvement in the efficiency and sensitivity of miRNA reverse transcription. The obtained cDNA can be directly used for qPCR detection using either SYBR Green dye-base or Taqman probe-base reagent.
Catalog No. 50-001-37775 Supplier ABclonal Technology Supplier No. RK3017020RXN
This kit is suitable for cDNA first strand synthesis using microRNA as template through the tail addition method, where the Poly (A) tail addition reaction and reverse transcription reaction at the 3 end of miRNA can be efficiently carried out simultaneously. ABScript miRNA-A Enzyme Mix contains Poly (A) Polymerase (PAP) and reverse transcriptase. PAP is mainly used to add Poly (A) tails at the 3 end of RNA molecules, and can also specifically recognize single stranded RNA, effectively avoiding RT reactions of pre-miRNA with double stranded or stem-loop structures. The modified reverse transcriptase lacks of RNase H activity and increases its affinity with RNA, resulting in a significant improvement in the efficiency and sensitivity of miRNA reverse transcription. The obtained cDNA can be directly used for qPCR detection using either SYBR Green dye-base or Taqman probe-base reagent.