Learn More
ABclonal Technology DL2000 plus DNA Marker
SDP
Catalog No. 5000138630
Encompass_Preferred
The DL2000 plus DNA Marker is a ready-to-use DNA Marker, which is pre-mixed with loading dye for direct gelloading. This product consists of 6 individual DNA fragments: 100 bp, 250 bp, 500 bp, 750 bp, 1000 bp and 2000 bp, derived from amixture of PCR products and specifically digested plasmid DNA. The 2000 bp and 750 bp fragment are present at increased intensity for easy identification.
Catalog No. 50-001-38630 Supplier ABclonal Technology Supplier No. RK301902.500ML
The DL2000 plus DNA Marker is a ready-to-use DNA Marker, which is pre-mixed with loading dye for direct gelloading. This product consists of 6 individual DNA fragments: 100 bp, 250 bp, 500 bp, 750 bp, 1000 bp and 2000 bp, derived from amixture of PCR products and specifically digested plasmid DNA. The 2000 bp and 750 bp fragment are present at increased intensity for easy identification.