Learn More
ABclonal Technology DL15000 DNA Marker
SDP
Catalog No. 5000135310
Encompass_Preferred
The DL15000 DNA Marker is a ready-to-use DNA Marker, which is pre-mixed with loading dye for direct gelloading. This product consists of 7 individual DNA fragments: 250 bp, 1000 bp, 2500 bp, 5000 bp, 7500 bp, 10000 bp and 15000 bp derived from amixture of PCR products and specifically digested plasmid DNA.
Catalog No. 50-001-35310 Supplier ABclonal Technology Supplier No. RK301922.500ML
The DL15000 DNA Marker is a ready-to-use DNA Marker, which is pre-mixed with loading dye for direct gelloading. This product consists of 7 individual DNA fragments: 250 bp, 1000 bp, 2500 bp, 5000 bp, 7500 bp, 10000 bp and 15000 bp derived from amixture of PCR products and specifically digested plasmid DNA.