Learn More
ABclonal Technology 100 bp DNA Ladder
SDP
Catalog No. 5000135018
Encompass_Preferred
The 100 bp DNA Ladder is a ready-to-use DNA Ladder, which is pre-mixed with loading dye for direct gelloading. This product consists of 11 individual DNA fragments: 100 bp, 200 bp, 300 bp, 400 bp, 500 bp, 600 bp, 700 bp, 800 bp, 900 bp, 1000 bp and 1500 bp, derived from amixture of PCR products and specifically digested plasmid DNA. The 500 bp fragment is present at increased intensity for easy identification.
Catalog No. 50-001-35018 Supplier ABclonal Technology Supplier No. RK301932.500ML
The 100 bp DNA Ladder is a ready-to-use DNA Ladder, which is pre-mixed with loading dye for direct gelloading. This product consists of 11 individual DNA fragments: 100 bp, 200 bp, 300 bp, 400 bp, 500 bp, 600 bp, 700 bp, 800 bp, 900 bp, 1000 bp and 1500 bp, derived from amixture of PCR products and specifically digested plasmid DNA. The 500 bp fragment is present at increased intensity for easy identification.