Learn More
ABclonal Technology DL1000 DNA Marker
SDP
Catalog No. 5000135040
Encompass_Preferred
The DL1000 DNA Marker is a ready-to-use DNA Marker, which is pre-mixed with loading dye for direct gelloading. This product consists of 8 individual DNA fragments: 1000 bp, 2000 bp, 3000 bp, 4000 bp, 5000 bp, 6000 bp, 8000 bp and 10000 bp, derived from amixture of PCR products and specifically digested plasmid DNA.
Catalog No. 50-001-35040 Supplier ABclonal Technology Supplier No. RK301942.500ML
The DL1000 DNA Marker is a ready-to-use DNA Marker, which is pre-mixed with loading dye for direct gelloading. This product consists of 8 individual DNA fragments: 1000 bp, 2000 bp, 3000 bp, 4000 bp, 5000 bp, 6000 bp, 8000 bp and 10000 bp, derived from amixture of PCR products and specifically digested plasmid DNA.