Learn More
ABclonal Technology AFTMag Plasmid Mini Kit V2
SDP
Catalog No. 5000136555
Encompass_Preferred
This kit is a nucleic acid extraction kit based on magnetic beads, primarily designed for isolating and purifying high-quality plasmid DNA from bacteria. It utilizes the functional groups on the surface of magnetic beads to enrich nucleic acids from sample lysates onto the surface of the magnetic beads, followed by separation of the magnetic beads using a magnetic separation device to achieve rapid isolation and purification of nucleic acids. This reagent kit boasts high efficiency, speed, and convenience, and when paired with a fully automatic nucleic acid extraction instrument, it can complete the extraction of 96 samples in just approximately 25 minutes. Additionally, the plasmid DNA extracted using this kit is of high purity with intact fragments, making it suitable for direct use in molecular biology experiments such as transformation, PCR amplification, sequencing, ligation, and transfection of conventional cells.
Catalog No. 50-001-36555 Supplier ABclonal Technology Supplier No. RK3021820RXN
This kit is a nucleic acid extraction kit based on magnetic beads, primarily designed for isolating and purifying high-quality plasmid DNA from bacteria. It utilizes the functional groups on the surface of magnetic beads to enrich nucleic acids from sample lysates onto the surface of the magnetic beads, followed by separation of the magnetic beads using a magnetic separation device to achieve rapid isolation and purification of nucleic acids. This reagent kit boasts high efficiency, speed, and convenience, and when paired with a fully automatic nucleic acid extraction instrument, it can complete the extraction of 96 samples in just approximately 25 minutes. Additionally, the plasmid DNA extracted using this kit is of high purity with intact fragments, making it suitable for direct use in molecular biology experiments such as transformation, PCR amplification, sequencing, ligation, and transfection of conventional cells.