Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rkh acetate | 99.8% | 439.51 | 5 MG
SDP

Catalog No. 5000423257
Encompass_Preferred

RKH acetate exerts protective effects against sepsis-induced death and organ damage. It directly binds to Toll-like receptor 4 (TLR4) and blocks TLR4 signal transduction in immune cells. It is for research use only.

  • Target: TLR4
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Arg-Lys-His
  • Solubility in DMSO: 200 mg/mL
  • Solubility in H2O: ≥ 30 mg/mL

Catalog No. 50-004-23257 Supplier Medchemexpress LLC Supplier No. HYP10208A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RKH acetate exerts protective effects against sepsis-induced death and organ damage. It directly binds to Toll-like receptor 4 (TLR4) and blocks TLR4 signal transduction in immune cells. It is for research use only.

  • Target: TLR4
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Arg-Lys-His
  • Solubility in DMSO: 200 mg/mL
  • Solubility in H2O: ≥ 30 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.