Learn More
Selleck Chemical LLC RKI-1447 10mg 1342278-01-6
SDP
Supplier:  Selleck Chemical LLC S719510MG
Encompass_Preferred
RKI-1447 is a potent inhibitor of ROCK1 and ROCK2, with IC50 of 14.5 nM and 6.2 nM, respectively, has anti-invasive and antitumor activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4509