Learn More
ATCC RKO; Colon Carcinoma; Human (Homo sapiens)
Catalog No. 502381720
Encompass
RKO is a poorly differentiated colon carcinoma cell line and is the parental cell line (isogenic) of RKO-E6 (ATCC CRL-2578) and RKO-A545-1 (ATCC CRL-2579). It can be used as the control cell line for investigating the effects of p53 and gadd45 on cellular parameters.
Catalog No. 50-238-1720 Supplier ATCC Supplier No. CRL2577
RKO is a poorly differentiated colon carcinoma cell line and is the parental cell line (isogenic) of RKO-E6 (ATCC CRL-2578) and RKO-A545-1 (ATCC CRL-2579). It can be used as the control cell line for investigating the effects of p53 and gadd45 on cellular parameters.