Learn More
Abcam RKRARKE, PKG Inhibitor, 5MG
Catalog No. 502705435
Encompass_Preferred
MW 943.12 g/mol, Purity >99%. Competitive protein kinase G (PKG) inhibitor. Ki = 86 µM for PKG; Ki = 550 µM for PKA. Potential inhibitory activity in neuronal systems.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5435 Supplier Abcam Supplier No. AB1450635MG
MW 943.12 g/mol, Purity >99%. Competitive protein kinase G (PKG) inhibitor. Ki = 86 µM for PKG; Ki = 550 µM for PKA. Potential inhibitory activity in neuronal systems.
The product is subject to the following: Abcam Restricted Use Statement