Learn More
Aviva Systems Biology Corp RLBP1L1 Antibody - middle region (ARP53265_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53265_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RLBP1L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RLBP1L1. Alternate names: CRALBPL, FLJ37248, MGC34646, RLBP1L1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60984