Learn More
Aviva Systems Biology Corp RLF Antibody - N-terminal region (ARP38997_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38997_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RLF. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RLF. Alternate names: MGC142226, ZN-15L, ZNF292LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Pig
Catalog No. 50-284-772