Learn More
Aviva Systems Biology Corp RLN1 Antibody - middle region (ARP56721_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56721_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RLN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RLN1. Alternate names: H1, RLXH1, bA12D24.3.1, bA12D24.3.2ApplicationWB. Species reactivity: Human, Pig, Rat
Catalog No. 50-286-203