Learn More
TARGETMOL CHEMICALS INC RLX-33 5MG
SDP
Catalog No. 502579452
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. RLX-33 is a potent and selective antagonist of relaxin family peptide 3 (RXFP3) that crosses the blood-brain barrier and also blocks relaxin 3-induced ERK1/2 phosphorylation. RLX-33 can block the increase in food intake induced by the RXFP3 selective agonist R3/I5 in rats.RLX-33 can be used to study the metabolic syndrome. purity: 99%
Catalog No. 50-257-9452 Supplier TARGETMOL CHEMICALS INC Supplier No. T629295MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. RLX-33 is a potent and selective antagonist of relaxin family peptide 3 (RXFP3) that crosses the blood-brain barrier and also blocks relaxin 3-induced ERK1/2 phosphorylation. RLX-33 can block the increase in food intake induced by the RXFP3 selective agonist R3/I5 in rats.RLX-33 can be used to study the metabolic syndrome. purity: 99%