Learn More
ABclonal Technology Agarose
SDP
Catalog No. 5000135016
Encompass_Preferred
ABclonal Agarose powder is highly purified, applicable for a variety of molecular biology applications. It can be used for analysis of nucleic acids of various sizes (from 50 bp to 15 kbp).
Catalog No. 50-001-35016 Supplier ABclonal Technology Supplier No. RM02852100G
ABclonal Agarose powder is highly purified, applicable for a variety of molecular biology applications. It can be used for analysis of nucleic acids of various sizes (from 50 bp to 15 kbp).