Learn More
ABclonal Technology RNase A (Powder)
SDP
Supplier:  ABclonal Technology RM21314100MG
Encompass_Preferred
Ribonuclease A (RNase A), molecular weight 13.7 kDa, is an endonuclease that specifically acts on the 3 end of the pyrimidine residue on RNA, cleaving phosphodiester bonds formed with adjacent nucleotides, and the final product of the reaction is pyrimidine 3 phosphate and an oligonucleotide with pyrimidine 3 phosphate at the end. RNase A does not work on DNA.This product is sourced from bovine pancreas and is supplied in the form of white lyophilized powder with enzyme activity ≥ 50 Kunitz units/mg protein. Its recommended working concentration is 1-100 μg/mL, which will vary depending on specific applications.
Catalog No. 50-001-38038