Learn More
ABclonal Technology DNase I (Powder)
SDP
Supplier:  ABclonal Technology RM298651G
Encompass_Preferred
Deoxyribonuclease I (DNase I) is an endonuclease that nonspecifically cleaves DNA to release di-, tri-, and oligonucleotide products with 5phosphorylated and 3?-hydroxylated ends. This product is sourced from bovine pancreas and is supplied in powder form with enzyme activity ≥ 500 Kunitz units/mg protein.
Catalog No. 50-001-38634