Learn More
ABclonal Technology DNase I (Powder)
SDP
Catalog No. 5000138635
Encompass_Preferred
Deoxyribonuclease I (DNase I) is an endonuclease that nonspecifically cleaves DNA to release di-, tri-, and oligonucleotide products with 5phosphorylated and 3?-hydroxylated ends. This product is sourced from bovine pancreas and is supplied in powder form with enzyme activity ≥ 500 Kunitz units/mg protein.
Catalog No. 50-001-38635 Supplier ABclonal Technology Supplier No. RM29865100MG
Deoxyribonuclease I (DNase I) is an endonuclease that nonspecifically cleaves DNA to release di-, tri-, and oligonucleotide products with 5phosphorylated and 3?-hydroxylated ends. This product is sourced from bovine pancreas and is supplied in powder form with enzyme activity ≥ 500 Kunitz units/mg protein.