Learn More
Medchemexpress LLC RMC-0331 (RM-023) | 2488788-52-7 | 98.0% | C22H25ClF3N5O3 | 10MG
SDP
Catalog No. 5000260053
Encompass_Preferred
RMC-0331 is a potent, selective, orally bioavailable SOS1 inhibitor used as an in vivo tool compound to block RAS activation by disrupting the RAS-SOS1 interaction. It is reported with CAS 2488788-52-7 and chemical formula C22H25ClF3N5O3, and is supplied as a solid for preclinical research use.
  • Potent and selective SOS1 inhibition for RAS pathway studies.
  • Orally bioavailable profile suitable for in vivo experiments.
  • Reported chemical formula C22H25ClF3N5O3 and CAS 2488788-52-7 for identification.
  • Typical reported purity greater than 98%; confirm batch certificate of analysis.
  • Provided as a solid with recommended storage at -20 °C.
Catalog No. 50-002-60053 Supplier Medchemexpress LLC Supplier No. HY13488510MG
RMC-0331 is a potent, selective, orally bioavailable SOS1 inhibitor used as an in vivo tool compound to block RAS activation by disrupting the RAS-SOS1 interaction. It is reported with CAS 2488788-52-7 and chemical formula C22H25ClF3N5O3, and is supplied as a solid for preclinical research use.
  • Potent and selective SOS1 inhibition for RAS pathway studies.
  • Orally bioavailable profile suitable for in vivo experiments.
  • Reported chemical formula C22H25ClF3N5O3 and CAS 2488788-52-7 for identification.
  • Typical reported purity greater than 98%; confirm batch certificate of analysis.
  • Provided as a solid with recommended storage at -20 °C.