Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RMC-4550 10 MM 1 ML IN DMSO
SDP

Supplier:  Medchemexpress LLC RMC4550

Encompass_Preferred

RMC-4550 10 mM 1 mL in DMSO. Purity 99.21% CAS No. 2172651-73-7

Catalog No. NC2411097


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.