Learn More
Selleck Chemical LLC RMC-4550 1mg 2172651-73-7
SDP
Supplier:  Selleck Chemical LLC S87181MG
Encompass_Preferred
RMC-4550 is a potent SHP-2inhibitor with an IC50 of 1.55 nM and it has no significant activity vs. 468 kinases, the catalytic domain of 15 phosphatases and other cellular targets including GPCRs, transporters and ion channels. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8115