Learn More
Medchemexpress LLC RMC-4550 25mg | 2172651-73-7 | 437.4 g·mol-1 | C21H26Cl2N4O2 | 25 MG
SDP
Catalog No. 5000229030
Encompass_Preferred
RMC-4550 is a small-molecule, allosteric inhibitor of SHP2 supplied as a solid powder for laboratory research and typically packaged in a 25 mg vial for preclinical use.
  • Allosteric SHP2 inhibitor for preclinical research.
  • Reported low nanomolar potency against full-length SHP2 in biochemical assays.
  • High purity suitable for biochemical and cell-based assays.
  • Stable when stored cold and desiccated for long-term storage.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-29030 Supplier Medchemexpress LLC Supplier No. HY11600925MG
RMC-4550 is a small-molecule, allosteric inhibitor of SHP2 supplied as a solid powder for laboratory research and typically packaged in a 25 mg vial for preclinical use.
  • Allosteric SHP2 inhibitor for preclinical research.
  • Reported low nanomolar potency against full-length SHP2 in biochemical assays.
  • High purity suitable for biochemical and cell-based assays.
  • Stable when stored cold and desiccated for long-term storage.
  • For research use only; not for human therapeutic use.