Learn More
Medchemexpress LLC HY-132168 1mg , RMC-5552 CAS:2382768-62-7 Purity:>98%
SDP
Catalog No. 502259241
Encompass_Preferred
HY-132168 1mg RMC-5552 CAS: 2382768-62-7 RMC-5552 is a potent and selective inhibitor of mTORC1. RMC-5552 inhibits phosphorylation of mTORC1 pS6K and p4EBP1 with IC50s of 0.14 nM and 0.48 nM, respectively. RMC-5552 has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9241 Supplier Medchemexpress LLC Supplier No. HY1321681MG
HY-132168 1mg RMC-5552 CAS: 2382768-62-7 RMC-5552 is a potent and selective inhibitor of mTORC1. RMC-5552 inhibits phosphorylation of mTORC1 pS6K and p4EBP1 with IC50s of 0.14 nM and 0.48 nM, respectively. RMC-5552 has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.