Learn More
Chemie Tek RMC-6236, 5mg
SDP
Supplier:  Chemie Tek CTRMC62365MG
Encompass_Preferred
RMC-6236 is a First-in-Class, non-covalent, oral, potent, and RAS-Selective Tri-Complex RASMULTI(ON) Inhibitor with activity across a diverse spectrum of RAS mutants, including KRASG12V and KRASG12D
Catalog No. NC2891325