Learn More
Chemie Tek RMC-9805 (Zoldonrasib) 5mg
SDP
Catalog No. NC3626732
Encompass_Preferred
RMC-9805 (Zoldonrasib) is a first-in-class, mutant-selective, covalent and orally bioavailable inhibitor of KRAS G12D (ON) inhibitor (refs. 1, 2).
Catalog No. NC3626732 Supplier Chemie Tek Supplier No. CTRMC980510MG
RMC-9805 (Zoldonrasib) is a first-in-class, mutant-selective, covalent and orally bioavailable inhibitor of KRAS G12D (ON) inhibitor (refs. 1, 2).