Learn More
Aviva Systems Biology Corp RMND1 Antibody - middle region (ARP57061_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57061_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RMND1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RMND1. Alternate names: C6orf96, FLJ20627, MGC117362, MGC149570, MGC88260, RMD1, bA351K16, bA351K16.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62069