Learn More
Eurofins Discoverx Recombinant Murine CCL22, 5 µg. (92-1016)
Supplier:  Eurofins Discoverx 921016
Encompass
Murine CCL22 is a small cytokine protein that induces the migration of CCR4+ Th2 cells, regulatory T cells, dendritic cells, and NK cells. CCL22 secretion is induced by LPS and CD40 antibody in macrophages, and is upregulated by Th2 cytokines (IL-4 and IL-5) and downregulated by Th1 cytokines (IFNγ). Carries out its functions primarily by interacting with its chemokine receptor, CCR4, as an agonist. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9565