Learn More
Eurofins Discoverx Recombinant Murine CCL24, 5 µg. (92-1017)
Supplier:  Eurofins Discoverx 921017
Encompass
Murine CCL24 is a small cytokine protein produced by activated monocytes and T lymphocytes which selectively chemoattracts cells expressing CCR3, including eosinophils, basophils, Th2 T cells, mast cells, and certain subsets of dendritic cells. CCL24 also inhibits the proliferation of multipotent hematopoietic progenitor cells. Acts as an agonist for CCR3. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9566