Learn More
Eurofins Discoverx Recombinant Murine CCL3, 2 µg. (92-1015)
Supplier:  Eurofins Discoverx 921015
Encompass
Murine CCL3 is a small cytokine protein that acts as an agonist for CCR1 and CCR5. It plays a critical role in the recruitment of leukocytes to the site of inflammation. While both CCL3 and CCL4 attract monocytes, macrophages, and dendritic cells to the site of infection, CCL3 attracts CD8+ T cells, while CD4+ T cells are more responsive to CCL4. In addition to these functions, CCL3 also induces inflammatory cytokine secretion, mast cell degranulation, and NK cell activation. Provided as 2 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9564