Learn More
Eurofins Discoverx Recombinant Murine CXCL16, 5 µg. (92-1021)
Supplier:  Eurofins Discoverx 921021
Encompass
Murine CXCL16 is a cytokine protein expressed as a cell surface bound molecule, as well as a soluble chemokine, by macrophages, monocytes, B cells, and Schwann cells. Its expression is induced by IFNG and TNF-α. The soluble protein is chemotactic for activated CD8 T cells, inducing the process of calcium flux and regulating their migration. CXCL16 is an agonist for the CXCR6 receptor, but also serves as a receptor for oxidized LDL and phosphatidylserine. Provided as 5 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9570