Learn More
Electron Microscopy Sciences Tough-Tag Direct Thermal Removable Labels, 1.05 x 0.50"
SDP
Supplier:  Electron Microscopy Sciences 7751703
Encompass_Preferred
Durable labels are easily removed from surfaces without leaving any adhesive. Use for tracking of samples from purification to processing. The labels are resistant to most laboratory solvents and have shown short-term resistance to Acetone and Xylene. Use for beakers, bottles, flasks, glass vials, tubes, tube racks and pipettes.
Size: 1.05 x 0.50"
Catalog No. 50-190-2690