Learn More
TARGETMOL CHEMICALS INC RN-1 dihydrochloride 10MG
SDP
Catalog No. 502876002
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. RN-1 dihydrochloride is an effective and selective irreversible inhibitor of lysine-specific demethylase 1 (LSD1, IC50 = 70 nM). Purity 99.61%
Catalog No. 50-287-6002 Supplier TARGETMOL CHEMICALS INC Supplier No. T2165210MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. RN-1 dihydrochloride is an effective and selective irreversible inhibitor of lysine-specific demethylase 1 (LSD1, IC50 = 70 nM). Purity 99.61%